Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
419090
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
2210018M11RIK, C11ORF30, C1H11orf30
Chromosome Location
Chr.1: 195122953 - 195162521 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025146941.1 XP_025002709.1 15 - 16 2603 106
GenBank mRNA BX934563.1 - 2 - 3 310 106

View Full Product Details