Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
416770
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
-
Chromosome Location
Chr.15: 678598 - 684711 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001232857.5 XP_001232858.1 2 - 3 425 76
GenBank mRNA AJ720874.1 - 2 - 3 393 76

View Full Product Details