Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL41
Entrez Gene ID
424164
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10
Chromosome Location
Chr.7: 18661166 - 18671704 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_422010.6 XP_422010.1 5 - 6 2404 98
GenBank mRNA CR405927.1 - 5 - 6 781 98

View Full Product Details