Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC35E3
Entrez Gene ID
417842
Gene Name
solute carrier family 35 member E3
Gene Aliases
-
Chromosome Location
Chr.1: 35393882 - 35399422 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199616.1 NP_001186545.1 1 - 2 506 66
GenBank mRNA CR406003.1 - 1 - 2 232 66
DR428244.1 - 1 - 2 506 66

View Full Product Details