Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35E3
Entrez Gene ID
417842
Gene Name
solute carrier family 35 member E3
Gene Aliases
-
Chromosome Location
Chr.1: 35393882 - 35399422 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199616.1 NP_001186545.1 4 - 5 848 110
GenBank mRNA BM488687.1 - 3 - 4 233 110
CR406003.1 - 4 - 5 574 110

View Full Product Details