Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM7
Entrez Gene ID
419760
Gene Name
RNA binding motif protein 7
Gene Aliases
-
Chromosome Location
Chr.24: 4592248 - 4595675 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006318.1 NP_001006318.1 2 - 3 302 118
XM_015297995.2 XP_015153481.1 2 - 3 332 118
GenBank mRNA AJ719791.1 - 2 - 3 302 118

View Full Product Details