Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX11G
Entrez Gene ID
420131
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
-
Chromosome Location
Chr.28: 4159781 - 4161924 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006340.1 NP_001006340.1 2 - 3 274 107
GenBank mRNA AJ720362.1 - 2 - 3 274 107

View Full Product Details