Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTERF3
Entrez Gene ID
420237
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
MTERFD1
Chromosome Location
Chr.2: 126722212 - 126738826 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006348.1 NP_001006348.1 3 - 4 798 129
GenBank mRNA AJ720506.1 - 3 - 4 798 129

View Full Product Details