Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRPEL1
Entrez Gene ID
422865
Gene Name
GrpE like 1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.4: 79934605 - 79941011 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006458.1 NP_001006458.1 2 - 3 253 75
GenBank mRNA AJ721028.1 - 2 - 3 253 75

View Full Product Details