Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF2S1
Entrez Gene ID
423279
Gene Name
eukaryotic translation initiation factor 2 subunit alpha
Gene Aliases
-
Chromosome Location
Chr.5: 29170736 - 29180567 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006477.1 NP_001006477.1 5 - 6 681 94
XM_025150723.1 XP_025006491.1 5 - 6 675 94
GenBank mRNA AJ719612.1 - 5 - 6 681 94

View Full Product Details