Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNIP4
Entrez Gene ID
395239
Gene Name
potassium voltage-gated channel interacting protein 4
Gene Aliases
KCHIP4.2
Chromosome Location
Chr.4: 74568444 - 74948433 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204555.1 NP_989886.1 2 - 3 188 132
XM_025149477.1 XP_025005245.1 3 - 4 389 132
XM_025149478.1 XP_025005246.1 3 - 4 429 132
XM_025149479.1 XP_025005247.1 3 - 4 371 132
XM_025149480.1 XP_025005248.1 2 - 3 650 132
XM_025149481.1 XP_025005249.1 2 - 3 382 132
XM_025149482.1 XP_025005250.1 2 - 3 339 132
XM_025149483.1 XP_025005251.1 2 - 3 409 132
XM_025149484.1 XP_025005252.1 2 - 3 286 132
XM_025149485.1 XP_025005253.1 2 - 3 730 132
XM_025149486.1 XP_025005254.1 2 - 3 443 132
XM_025149487.1 XP_025005255.1 2 - 3 245 132
GenBank mRNA AF508737.1 - 2 - 3 188 132
DQ099910.1 - 2 - 3 184 132

View Full Product Details