Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRF9
Entrez Gene ID
395243
Gene Name
interferon regulatory factor 9
Gene Aliases
IRF10
Chromosome Location
Chr.20: 10063697 - 10066828 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204558.1 NP_989889.1 5 - 6 656 145
XM_025142159.1 XP_024997927.1 5 - 6 957 145
GenBank mRNA AF380350.1 - 5 - 6 656 145

View Full Product Details