Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPZ
Entrez Gene ID
395266
Gene Name
carboxypeptidase Z
Gene Aliases
-
Chromosome Location
Chr.4: 81163348 - 81197855 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204578.4 NP_989909.3 1 - 2 174 72
GenBank mRNA AF351205.1 - 1 - 2 72

View Full Product Details