Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARK7
Entrez Gene ID
395277
Gene Name
Parkinsonism associated deglycase
Gene Aliases
DJ-1
Chromosome Location
Chr.21: 264288 - 272660 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204585.1 NP_989916.1 6 - 7 551 97
XM_015296819.2 XP_015152305.1 6 - 7 490 97
GenBank mRNA AB076264.1 - 6 - 7 551 97
HM012714.1 - 5 - 6 430 97

View Full Product Details