Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPN
Entrez Gene ID
395298
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
SNRB', SNRPB, SNRPB', SNRPL
Chromosome Location
Chr.20: 10750447 - 10753198 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204599.1 NP_989930.1 6 - 7 760 88
GenBank mRNA AF134830.1 - 6 - 7 760 88
CR385829.1 - 6 - 7 740 88

View Full Product Details