Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPG5
Entrez Gene ID
395466
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
CALEB
Chromosome Location
Chr.2: 901382 - 910800 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204719.1 NP_990050.1 3 - 4 1887 92
GenBank mRNA Y09264.1 - - 1887 92

View Full Product Details