Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNM
Entrez Gene ID
395599
Gene Name
synemin
Gene Aliases
DMN
Chromosome Location
Chr.10: 17303647 - 17324081 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204809.4 NP_990140.2 3 - 4 1051 140

View Full Product Details