Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOLR1
Entrez Gene ID
395638
Gene Name
folate receptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 197315894 - 197319365 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204834.1 NP_990165.1 3 - 4 1249 51
XM_015280910.2 XP_015136396.1 4 - 5 652 51
GenBank mRNA AF160960.1 - 3 - 4 1249 51

View Full Product Details