Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TENM1
Entrez Gene ID
395668
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, TEN-1
Chromosome Location
Chr.4: 15205742 - 15529114 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204862.1 NP_990193.1 3 - 4 717 69
XM_015278189.2 XP_015133675.1 4 - 5 69
XM_015278190.2 XP_015133676.1 4 - 5 69
XM_015278191.2 XP_015133677.1 4 - 5 69
GenBank mRNA AJ238613.1 - 3 - 4 717 69

View Full Product Details