Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMX1B
Entrez Gene ID
396312
Gene Name
LIM homeobox transcription factor 1 beta
Gene Aliases
LMX, LMX1
Chromosome Location
Chr.17: 10357033 - 10450700 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205358.1 NP_990689.2 2 - 3 325 104
GenBank mRNA L39880.1 - 2 - 3 325 104
U41823.1 - 2 - 3 250 104

View Full Product Details