Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYP
Entrez Gene ID
396359
Gene Name
synaptophysin
Gene Aliases
SYNPR
Chromosome Location
Chr.12: 13810269 - 13889947 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205394.1 NP_990725.1 3 - 4 427 80
XM_015293015.1 XP_015148501.1 4 - 5 575 80
XM_015293016.2 XP_015148502.1 3 - 4 518 80
GenBank mRNA BX936122.2 - 3 - 4 379 80
U27708.1 - 3 - 4 427 80

View Full Product Details