Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRF1
Entrez Gene ID
396384
Gene Name
interferon regulatory factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 17591208 - 17597351 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205415.1 NP_990746.1 7 - 8 797 136
XM_015294014.1 XP_015149500.1 7 - 8 992 136
XM_015294015.1 XP_015149501.1 7 - 8 992 136
GenBank mRNA KC250010.1 - 6 - 7 677 136
L39766.1 - 7 - 8 797 136

View Full Product Details