Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKLR
Entrez Gene ID
396456
Gene Name
pyruvate kinase, liver and RBC
Gene Aliases
PKM, PKM2
Chromosome Location
Chr.10: 2035711 - 2052967 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205469.1 NP_990800.1 2 - 3 233 67
XM_015278795.2 XP_015134281.1 2 - 3 235 67
XM_015278796.2 XP_015134282.1 3 - 4 295 67
XM_025153808.1 XP_025009576.1 3 - 4 295 67
GenBank mRNA J00903.1 - 2 - 3 233 67

View Full Product Details