Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKLR
Entrez Gene ID
396456
Gene Name
pyruvate kinase, liver and RBC
Gene Aliases
PKM, PKM2
Chromosome Location
Chr.10: 2035711 - 2052967 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205469.1 NP_990800.1 4 - 5 454 75
XM_015278795.2 XP_015134281.1 4 - 5 456 75
XM_015278796.2 XP_015134282.1 5 - 6 516 75
XM_025153808.1 XP_025009576.1 5 - 6 516 75
GenBank mRNA J00903.1 - 4 - 5 454 75

View Full Product Details