Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB5A
Entrez Gene ID
420649
Gene Name
RAB5A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.2: 35764554 - 35781507 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006363.2 NP_001006363.1 5 - 6 892 129
XM_015281490.2 XP_015136976.1 5 - 6 792 129
GenBank mRNA AJ720737.1 - 5 - 6 893 129

View Full Product Details