Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSR1
Entrez Gene ID
420871
Gene Name
signal sequence receptor subunit 1
Gene Aliases
-
Chromosome Location
Chr.2: 64612806 - 64647744 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006375.1 NP_001006375.1 6 - 7 787 126
XM_015275890.2 XP_015131376.1 10 - 11 2746 126
XM_015275891.2 XP_015131377.1 10 - 11 2746 126
GenBank mRNA AJ719521.1 - 6 - 7 787 126

View Full Product Details