Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3L
Entrez Gene ID
418033
Gene Name
eukaryotic translation initiation factor 3 subunit L
Gene Aliases
EIF3EIP, EIF3S6IP
Chromosome Location
Chr.1: 51097089 - 51106702 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012535.2 NP_001012553.1 6 - 7 541 146
GenBank mRNA AJ851472.1 - 6 - 7 542 146

View Full Product Details