Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMG1
Entrez Gene ID
418520
Gene Name
proteasome assembly chaperone 1
Gene Aliases
DSCR2
Chromosome Location
Chr.1: 109260957 - 109269886 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012543.1 NP_001012561.1 4 - 5 469 112
GenBank mRNA AJ851676.1 - 4 - 5 469 112
BX931893.1 - 3 - 4 250 109
BX950532.1 - 3 - 4 523 112

View Full Product Details