Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RELL1
Entrez Gene ID
422795
Gene Name
RELT like 1
Gene Aliases
-
Chromosome Location
Chr.4: 69834800 - 69855846 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012582.1 NP_001012600.1 3 - 4 461 94
GenBank mRNA AJ851746.1 - 3 - 4 461 94
CR523601.1 - 3 - 4 442 94

View Full Product Details