Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC62
Entrez Gene ID
424993
Gene Name
SEC62 homolog, preprotein translocation factor
Gene Aliases
TLOC1
Chromosome Location
Chr.9: 20031455 - 20047148 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012602.1 NP_001012620.1 6 - 7 612 85
GenBank mRNA AJ851749.1 - 6 - 7 612 85

View Full Product Details