Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXL2
Entrez Gene ID
503512
Gene Name
forkhead box L2
Gene Aliases
-
Chromosome Location
Chr.9: 5874581 - 5875710 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012612.1 NP_001012630.1 1 - 1 554 86
GenBank mRNA AY487165.1 - 1 - 1 554 86
JF708868.1 - 1 - 1 554 86

View Full Product Details