Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIPIN
Entrez Gene ID
415548
Gene Name
TIMELESS interacting protein
Gene Aliases
-
Chromosome Location
Chr.10: 18896129 - 18905363 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012690.1 NP_001012708.1 6 - 7 650 105
XM_015292095.2 XP_015147581.1 6 - 7 975 105
GenBank mRNA AJ851484.1 - 6 - 7 650 105

View Full Product Details