Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM18
Entrez Gene ID
421911
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.3: 92123500 - 92128187 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012698.1 NP_001012716.1 2 - 3 205 112
GenBank mRNA AJ851490.1 - 2 - 3 205 112
CR353979.1 - 2 - 3 226 112
CR406940.1 - 2 - 3 185 112

View Full Product Details