Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFAP3
Entrez Gene ID
416261
Gene Name
microfibrillar associated protein 3
Gene Aliases
-
Chromosome Location
Chr.13: 12969677 - 12979688 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012784.2 NP_001012802.1 2 - 3 496 85
XM_015293781.2 XP_015149267.1 2 - 3 583 85
GenBank mRNA AJ851544.1 - 2 - 3 497 85
CR353656.1 - 2 - 3 512 85

View Full Product Details