Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
420806
Gene Name
ribosome binding factor A (putative)
Gene Aliases
C18orf22
Chromosome Location
Chr.2: 56089456 - 56105012 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012861.1 NP_001012879.1 4 - 5 520 149
XM_015275856.2 XP_015131342.1 4 - 5 1006 149
XM_015275857.2 XP_015131343.1 4 - 5 587 149
XM_015275858.2 XP_015131344.1 3 - 4 829 149
XM_015275859.2 XP_015131345.1 3 - 4 805 149
GenBank mRNA AJ719502.1 - 4 - 5 520 149
CR338760.1 - 4 - 5 495 149

View Full Product Details