Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL14
Entrez Gene ID
421100
Gene Name
kelch like family member 14
Gene Aliases
-
Chromosome Location
Chr.2: 106576890 - 106637507 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012866.1 NP_001012884.1 4 - 5 1407 67
GenBank mRNA AJ851615.1 - 4 - 5 1407 67

View Full Product Details