Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PELI3
Entrez Gene ID
421279
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases
PELI1
Chromosome Location
Chr.3: 10247152 - 10289373 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012872.2 NP_001012890.1 3 - 4 440 95
XM_015278047.2 XP_015133533.1 4 - 5 652 95
XM_015278048.2 XP_015133534.1 4 - 5 649 95
XM_015278049.2 XP_015133535.1 3 - 4 491 95
XM_015278050.2 XP_015133536.1 3 - 4 436 95
XM_015278051.2 XP_015133537.1 3 - 4 436 95
XM_025148641.1 XP_025004409.1 3 - 4 296 95
XM_025148642.1 XP_025004410.1 3 - 4 458 95
GenBank mRNA AJ719997.1 - 3 - 4 441 95

View Full Product Details