Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPNT1
Entrez Gene ID
421347
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
-
Chromosome Location
Chr.3: 18912027 - 18922847 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012874.2 NP_001012892.2 5 - 6 661 86
XM_015283646.2 XP_015139132.1 4 - 5 416 86
XM_015283647.2 XP_015139133.1 5 - 6 601 86
XM_015283648.2 XP_015139134.1 4 - 5 504 86
GenBank mRNA BX930248.2 - 4 - 5 396 86
BX931514.1 - 5 - 6 616 86
BX935292.2 - 4 - 5 406 86
CR385312.1 - 4 - 5 407 86

View Full Product Details