Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPNT1
Entrez Gene ID
421347
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
-
Chromosome Location
Chr.3: 18912027 - 18922847 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012874.2 NP_001012892.2 9 - 10 1099 107
XM_015283646.2 XP_015139132.1 8 - 9 854 107
XM_015283647.2 XP_015139133.1 9 - 10 1039 107
XM_015283648.2 XP_015139134.1 8 - 9 942 107

View Full Product Details