Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCCPDH
Entrez Gene ID
421485
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
-
Chromosome Location
Chr.3: 33951859 - 33961850 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012875.1 NP_001012893.1 2 - 3 366 64
GenBank mRNA AJ719381.1 - 2 - 3 366 64
BX933039.2 - 2 - 3 295 64
BX934370.1 - 2 - 3 295 64
CR353828.1 - 2 - 3 331 64

View Full Product Details