Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCCPDH
Entrez Gene ID
421485
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
-
Chromosome Location
Chr.3: 33951859 - 33961850 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012875.1 NP_001012893.1 5 - 6 632 75
GenBank mRNA AJ719381.1 - 5 - 6 632 75
BX933039.2 - 5 - 6 561 75
BX934370.1 - 5 - 6 561 75
CR353828.1 - 5 - 6 597 75

View Full Product Details