Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEURL1
Entrez Gene ID
423875
Gene Name
neuralized E3 ubiquitin protein ligase 1
Gene Aliases
NEURL
Chromosome Location
Chr.6: 24618495 - 24764948 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012910.1 NP_001012928.1 2 - 3 361 77
XM_015288647.2 XP_015144133.1 2 - 3 496 77
XM_015288648.2 XP_015144134.1 2 - 3 383 77
XM_025151610.1 XP_025007378.1 2 - 3 426 77
GenBank mRNA AJ719833.1 - 2 - 3 361 77

View Full Product Details