Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC40A1
Entrez Gene ID
423984
Gene Name
solute carrier family 40 member 1
Gene Aliases
-
Chromosome Location
Chr.7: 388104 - 402908 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012913.1 NP_001012931.1 5 - 6 728 97
XM_015289163.2 XP_015144649.1 6 - 7 825 97
GenBank mRNA AJ851647.1 - 5 - 6 728 97

View Full Product Details