Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDELR3
Entrez Gene ID
418026
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Gene Aliases
-
Chromosome Location
Chr.1: 50897173 - 50902281 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012996.1 NP_001013014.1 1 - 2 350 88
GenBank mRNA BX935103.2 - 1 - 2 158 88

View Full Product Details