Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASS1
Entrez Gene ID
417185
Gene Name
argininosuccinate synthase 1
Gene Aliases
-
Chromosome Location
Chr.17: 6181029 - 6201576 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013395.1 NP_001013413.1 5 - 6 498 83
GenBank mRNA AJ720611.1 - 5 - 6 498 83
BX930222.1 - 5 - 6 494 83
CR407399.1 - 5 - 6 645 83

View Full Product Details