Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTX3
Entrez Gene ID
548626
Gene Name
pentraxin 3
Gene Aliases
PPTX
Chromosome Location
Chr.9: 22869367 - 22874339 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017413.1 NP_001017413.1 1 - 2 424 61
GenBank mRNA AY672618.1 - 1 - 2 424 61
BX950436.2 - 1 - 2 262 61

View Full Product Details