Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF15
Entrez Gene ID
417247
Gene Name
tumor necrosis factor superfamily member 15
Gene Aliases
-
Chromosome Location
Chr.17: 2929784 - 2943040 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024578.1 NP_001019749.1 1 - 2 217 78
GenBank mRNA AB194710.1 - 1 - 2 217 78
BX930081.1 - 1 - 2 222 78
CR353356.1 - 2 - 3 325 78

View Full Product Details