Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCT
Entrez Gene ID
423015
Gene Name
secretin
Gene Aliases
-
Chromosome Location
Chr.5: 799459 - 818029 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024833.2 NP_001020004.2 3 - 4 197 81
GenBank mRNA AY954510.1 - 1 - 2 81
CR352860.1 - 3 - 4 196 81
EU686392.1 - 3 - 4 195 81

View Full Product Details