Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF18
Entrez Gene ID
419424
Gene Name
TNF receptor superfamily member 18
Gene Aliases
GITR
Chromosome Location
Chr.21: 2563494 - 2569804 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030874.1 NP_001026045.1 3 - 4 528 94
GenBank mRNA AJ719323.1 - 3 - 4 528 94
MH492023.1 - 3 - 4 387 94

View Full Product Details