Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID5B
Entrez Gene ID
423661
Gene Name
AT-rich interaction domain 5B
Gene Aliases
-
Chromosome Location
Chr.6: 8969664 - 9115288 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031220.1 NP_001026391.1 9 - 10 1413 125
XM_015288120.2 XP_015143606.1 8 - 9 1471 125
XM_015288121.2 XP_015143607.1 6 - 7 1213 125
XM_015288128.2 XP_015143614.1 16 - 17 5730 125
GenBank mRNA AJ720565.1 - 9 - 10 1413 125

View Full Product Details